Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Homophthalic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(Carboxymethyl)benzoic acid

12084-0250 12084-1000
TS12084-0250EA 43 USD InStock
TS12084-0250 TS12084-1000
Homophthalic acid 98%
Homophthalic acid


Formula: C₉H₈O₄
MW: 180.16 g/mol
Melting Pt: 178…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00004326
CAS Number: 89-51-0
EINECS: 201-913-9
Order Now
Specification Test Results
Appearance Off-white to light yellow Powder
Infrared spectrum Conforms
Melting point 178 to 182 °C
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty