Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Iodobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: p-Iodobenzoic acid

12448-0500 12448-0100
TS12448-0500EA 203 USD InStock
TS12448-0500 TS12448-0100
4-Iodobenzoic acid 98%
4-Iodobenzoic acid


Formula: C₇H₅IO₂
MW: 248.02 g/mol
Boiling Pt: 319 °C (1013 hPa)
Melting Pt: 270…274 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002533
CAS Number: 619-58-9
EINECS: 210-603-2
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 270 to 274 °C
HPLC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty