Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Terephthalaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Terephthaldicarboxaldehydep-PhthalaldehydeTerephthaldialdehyde

13789-1000 13789-5000
TS13789-5000EA 225 USD InStock
TS13789-5000 TS13789-1000
Terephthalaldehyde 98%
Terephthalaldehyde


Formula: C₈H₆O₂
MW: 134.13 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 114…116 °C
Flash Pt: 76 °C
MDL Number: MFCD00006949
CAS Number: 623-27-8
EINECS: 210-784-8
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 114 to 116 °C
GC ≥97.5 %
Specific optical rotation +35° to +39° (20 °C, 589 nm) (C=5, 1N HCl)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty