Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

o-Toluic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Methylbenzoic acid

13904-0050 13904-1000 13904-0010
TS13904-0050EA 26 USD InStock
TS13904-0050 TS13904-0010 TS13904-1000
o-Toluic acid 98+%
o-Toluic acid


Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 103…105 °C
Flash Pt: 148 °C
Storage Temperature: Ambient
MDL Number: MFCD00002477
CAS Number: 118-90-1
EINECS: 204-284-9
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder or needles
Infrared spectrum Conforms
Melting point 102 to 106 °C
Titration with NaOH ≥98.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty