Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(3,4,5-Trimethoxyphenyl)acetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3,4,5-Trimethoxyphenylacetic acid

13999-0500 13999-0050 13999-0100
TS13999-0500EA 253 USD InStock
TS13999-0500 TS13999-0100 TS13999-0050
(3,4,5-Trimethoxyphenyl)acetic acid 99%
(3,4,5-Trimethoxyphenyl)acetic acid


Formula: (C₆H₂(OCH₃)₃)CH₂COOH
MW: 226.23 g/mol
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 117…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00004336
CAS Number: 951-82-6
EINECS: 213-456-2
Order Now
Specification Test Results
Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 117 to 120 °C
Titration with NaOH ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty