Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trityl bromide 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: BromotriphenylmethaneTriphenylbromomethaneTriphenylmethyl bromide

14039-0500 14039-0100
TS14039-0100EA 37.7 USD InStock
TS14039-0100 TS14039-0500
Trityl bromide 98%
Trityl bromide


Formula: (C₆H₅)₃CBr
MW: 323.23 g/mol
Boiling Pt: 230 °C (15 mmHg)
Melting Pt: 151…155 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000120
CAS Number: 596-43-0
EINECS: 209-884-4
UN: 1759
ADR: 8,II
Order Now
Specification Test Results
Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 151 to 155 °C
Titration Argentometric ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty