Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Chloro-4-iodobenzene 99%

Supplier: Thermo Scientific Chemicals
Warning

14648-1000 14648-0250
TS14648-0250EA 47 USD InStock
TS14648-0250 TS14648-1000
1-Chloro-4-iodobenzene 99%
1-Chloro-4-iodobenzene


Formula: C₆H₄ClI
MW: 238.45 g/mol
Boiling Pt: 226 °C (1013 hPa)
Melting Pt: 52…55 °C
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00001053
CAS Number: 637-87-6
EINECS: 211-305-5
Order Now
Specification Test Results
Appearance Off-white to light beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 52 to 55 °C
GC ≥98.5 %
Heavy metals (as Pb) ≤10 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty