Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitroanisole 99+%

Supplier: Thermo Scientific Chemicals
Warning

14702-0050 14702-2500
TS14702-0050EA 27.16 USD InStock
TS14702-0050 TS14702-2500
4-Nitroanisole 99+%
4-Nitroanisole


Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 51…53 °C
Density: 1.23 g/cm³
Flash Pt: 130 °C
MDL Number: MFCD00007327
CAS Number: 100-17-4
EINECS: 202-825-3
UN: 3458
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Green to brown Crystals
Infrared spectrum Conforms
Melting point 51 to 53 °C
GC ≥99.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty