Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4,4'-Difluorobenzophenone 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Bis(4-Fluorophenyl)methanoneBis(4-fluorophenyl) ketone

15179-0050 15179-1000 15179-0250
TS15179-1000EA 280 USD InStock
TS15179-1000 TS15179-0250 TS15179-0050
4,4'-Difluorobenzophenone 99%
4,4'-Difluorobenzophenone


Formula: C₁₃H₈F₂O
MW: 218.2 g/mol
Boiling Pt: 200 °C (760 mmHg)
Melting Pt: 106…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00000353
CAS Number: 345-92-6
EINECS: 206-466-3
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 106 to 109 °C
Loss on drying ≤0.25 %
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty