Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Tolualdehyde 98% stabilized

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: o-Tolualdehyde2-Methylbenzaldehyde

15243-1000 15243-2500 15243-0250
TS15243-2500EA 344.05 USD InStock
TS15243-2500 TS15243-0250 TS15243-1000
2-Tolualdehyde 98% stabilized
2-Tolualdehyde


Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 199…200 °C (760 mmHg)
Melting Pt: –35 °C
Density: 1.039 g/cm³
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00003338
CAS Number: 529-20-4
EINECS: 208-452-2
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5460 to 1.5490 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer 0.1 to 0.5 % hydroquinone

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty