Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Aminobenzyl alcohol 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(Hydroxymethyl)aniline(2-Aminophenyl)methanol

15373-0250 15373-1000 15373-0100
TS15373-1000EA 177 USD InStock
TS15373-1000 TS15373-0100 TS15373-0250
2-Aminobenzyl alcohol 98%
2-Aminobenzyl alcohol


Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 162 °C (15 mmHg)
Melting Pt: 82…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00007749
CAS Number: 5344-90-1
EINECS: 226-293-7
Order Now
Specification Test Results
Appearance Beige to grey-brown Crystalline powder
Infrared spectrum Conforms
Melting point 82 to 86 °C
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty