Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-3-chlorobenzene 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Bromochlorobenzene

15420-1000 15420-0250
TS15420-1000EA 88 USD InStock
TS15420-1000 TS15420-0250
1-Bromo-3-chlorobenzene 99%
1-Bromo-3-chlorobenzene


Formula: C₆H₄BrCl
MW: 191.45 g/mol
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: –21.5 °C
Density: 1.63 g/cm³
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00000568
CAS Number: 108-37-2
EINECS: 203-575-8
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5760 to 1.5780 (20°C, 589 nm)
GC ≥98.5 %
NMR (1H-NMR) Consistent with structure

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty