Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chlorobenzaldehyde 99%

Supplier: Thermo Scientific Chemicals
Danger

15461-0050 15461-5000 15461-0010 15461-1000 15461-2500
TS15461-2500EA 57 USD InStock
TS15461-2500 TS15461-0010 TS15461-1000 TS15461-0050 TS15461-5000
2-Chlorobenzaldehyde 99%
2-Chlorobenzaldehyde


Formula: C₇H₅ClO
MW: 140.57 g/mol
Boiling Pt: 211.9 °C (1013 hPa)
Melting Pt: 10…11.5 °C
Density: 1.25 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00003304
CAS Number: 89-98-5
EINECS: 201-956-3
UN: 3265
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5650 to 1.5670 (20°C, 589 nm)
GC ≥98.5 %
Free acid ≤1 % (as chlorobenzoic acid)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty