Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2-Difluorobenzene 98%

Supplier: Thermo Scientific Chemicals
Danger

15515-0100 15515-0500
TS15515-0100EA 48 USD InStock
TS15515-0100 TS15515-0500
1,2-Difluorobenzene 98%
1,2-Difluorobenzene


Formula: C₆H₄F₂
MW: 114.09 g/mol
Boiling Pt: 92 °C (1013 hPa)
Melting Pt: –34 °C
Density: 1.158 g/cm³
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00000284
CAS Number: 367-11-3
EINECS: 206-680-7
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4417 to 1.4437 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty