Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methylbenzyl alcohol 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: p-Tolylmethanol4-(Hydroxymethyl)toluole(4-Methylphenyl)methanol

15592-0250 15592-0050 15592-1000
TS15592-0250EA 53.66 USD InStock
TS15592-0250 TS15592-0050 TS15592-1000
4-Methylbenzyl alcohol 98%
4-Methylbenzyl alcohol


Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 217 °C (760 mmHg)
Melting Pt: 56…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00004664
CAS Number: 589-18-4
EINECS: 209-639-1
Order Now
Specification Test Results
Appearance White Crystals or needles or crystalline mass
Infrared spectrum Conforms
Melting point 56 to 61 °C
GC ≥97.5 %
UV at 320 nm A: ≤0.1
UV at 350 nm A: ≤0.05
UV at 380 nm A: ≤0.01
UV at 400 nm A: ≤0.01
Color scale ≤15 APHA
Residue after evaporation ≤0.0005 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty