Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4,4'-Isopropylidenediphenol 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Bisphenol A2,2-Bis(4-hydroxyphenyl)propaneBPA

15824-0050 15824-1000 15824-5000 15824-0020
TS15824-5000EA 35 USD InStock
TS15824-5000 TS15824-1000 TS15824-0020 TS15824-0050
4,4'-Isopropylidenediphenol 97%
4,4'-Isopropylidenediphenol


Formula: C₁₅H₁₆O₂
MW: 228.29 g/mol
Boiling Pt: 220 °C (4 mmHg)
Melting Pt: 155…158 °C
Flash Pt: 227 °C
Storage Temperature: Ambient
MDL Number: MFCD00002366
CAS Number: 80-05-7
EINECS: 201-245-8
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Appearance White Flakes or prills
Infrared spectrum Conforms
Melting point 155 to 160 °C
GC ≥96.0 %
GC ≥98.5 % Mixture of isomers + Ethylbenzene
GC ≤25 % Ethylbenzene
GC ≤0.1 % Benzene
GC ≤0.4 % Toluene
Color scale ≤10 APHA
Residue after evaporation ≤0.002 %
Acidity/alkalinity ≤0.00025 meq/g
Aluminium (Al) ≤0.5 ppm
Cadmium (Cd) ≤0.1 ppm
Chromium (Cr) ≤0.05 ppm
Cobalt (Co) ≤0.1 ppm
Copper (Cu) ≤0.05 ppm
Iron (Fe) ≤0.2 ppm
Nickel (Ni) ≤0.05 ppm
Heavy metals (as Pb) ≤5 ppm
Sulfur compounds (as S) ≤0.003 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty