Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Acetylnaphthalene 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Methyl β-naphthyl ketone2'-Acetonaphthone2´-Acetonaphthone2-Acetonaphthone1-(Naphthalen-2-yl)ethanone

15888-1000 15888-5000
TS15888-5000EA 157.74 USD InStock
TS15888-5000 TS15888-1000
2-Acetylnaphthalene 99%
2-Acetylnaphthalene


Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 53…56 °C
Flash Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00004108
CAS Number: 93-08-3
EINECS: 202-216-2
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 53 to 56 °C
GC ≥98.5 %
Color scale ≤20 APHA
Ash ≤0.005 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty