Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4,6-Tribromophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

16010-5000 16010-0050 16010-1000
TS16010-1000EA 28.53 USD InStock
TS16010-1000 TS16010-0050 TS16010-5000
2,4,6-Tribromophenol 98%
2,4,6-Tribromophenol


Formula: C₆H₃Br₃O
MW: 330.8 g/mol
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 89…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00002150
CAS Number: 118-79-6
EINECS: 204-278-6
UN: 3077
Order Now
Specification Test Results
Appearance White to pink to light brown Powder or flakes and/or chunks
Infrared spectrum Conforms
Melting point 89 to 94 °C
Titration with NaOH ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty