Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3,5-Tribromobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

16108-1000 16108-0250
TS16108-1000EA 196 USD InStock
TS16108-1000 TS16108-0250
1,3,5-Tribromobenzene 98%
1,3,5-Tribromobenzene


Formula: C₆H₃Br₃
MW: 314.8 g/mol
Boiling Pt: 271 °C (1013 hPa)
Melting Pt: 118…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00000080
CAS Number: 626-39-1
EINECS: 210-947-3
Order Now
Specification Test Results
Appearance White to orange-brown Powder
Infrared spectrum Conforms
Melting point 118 to 123 °C
GC ≥97.5 %
Color scale ≤100 APHA

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty