Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Chloro-4-nitrotoluene 99%

Supplier: Thermo Scientific Chemicals
Danger

16109-2500 16109-0100 16109-1000
TS16109-2500EA 264 USD InStock
TS16109-2500 TS16109-1000 TS16109-0100
α-Chloro-4-nitrotoluene 99%
α-Chloro-4-nitrotoluene


Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 112 °C (.6 mmHg)
Melting Pt: 71…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00007374
CAS Number: 100-14-1
EINECS: 202-822-7
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance Light yellow Crystalline powder or needles
Infrared spectrum Conforms
Melting point 71 to 74 °C
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty