Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloroanisole 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Chloro-4-methoxybenzene

17015-0250 17015-1000
TS17015-1000EA 158 USD InStock
TS17015-1000 TS17015-0250
4-Chloroanisole 99%
4-Chloroanisole


Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 198…202 °C (760 mmHg)
Melting Pt: –18 °C
Density: 1.16 g/cm³
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00000616
CAS Number: 623-12-1
EINECS: 210-772-2
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥98.5 %
Specific optical rotation +220° to +235° (20°C, 589 nm) (c=1, H2O)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty