Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Chloro-m-cresol 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1-Chloro-2-hydroxy-4-methylbenzene2-Chloro-5-methylphenol

17041-0250 17041-1000 17041-5000
TS17041-1000EA 142 USD InStock
TS17041-1000 TS17041-0250 TS17041-5000
6-Chloro-m-cresol 99%
6-Chloro-m-cresol


Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 196 °C (760 mmHg)
Melting Pt: 45…48 °C
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00002175
CAS Number: 615-74-7
EINECS: 210-444-9
UN: 3437
ADR: 6.1,II
Order Now
Specification Test Results
Appearance White to yellow to beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 45 to 48 °C
GC ≥98.5 %
HPLC ≥88 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty