Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluorobenzylamine 96%

Supplier: Thermo Scientific Chemicals
Danger

17194-0100 17194-0500 17194-0010
TS17194-0500EA 319 USD InStock
TS17194-0500 TS17194-0010 TS17194-0100
2-Fluorobenzylamine 96%
2-Fluorobenzylamine


Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 73…75 °C (13 mmHg)
Density: 1.09 g/cm³
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00008107
CAS Number: 89-99-6
EINECS: 201-957-9
UN: 2735
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear colorless to orange Liquid
Infrared spectrum Conforms
Refractive index 1.5150 to 1.5190 (20°C, 589 nm)
GC ≥95 %
HPLC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty