Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluorobenzylamine 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (4-Fluorophenyl)methanamine

17195-1000 17195-0250
TS17195-0250EA 66.94 USD InStock
TS17195-0250 TS17195-1000
4-Fluorobenzylamine 97%
4-Fluorobenzylamine


Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 183…185 °C (760 mmHg)
Density: 1.09 g/cm³
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00008120
CAS Number: 140-75-0
EINECS: 205-430-4
UN: 2735
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5115 to 1.5155 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty