Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(Hydroxymethyl)benzyl alcohol 97%

Supplier: Thermo Scientific Chemicals
Synonyms: o-Phenylenedimethanol1,2-Benzenedimethanolo-Xylene-α,α'-diol

18041-0025 18041-0100
TS18041-0100EA 239 USD InStock
TS18041-0100 TS18041-0025
2-(Hydroxymethyl)benzyl alcohol 97%
2-(Hydroxymethyl)benzyl alcohol


Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 291 °C (760 mmHg)
Melting Pt: 61…64 °C
Flash Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00004626
CAS Number: 612-14-6
EINECS: 210-293-9
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 61.0 to 65.0 °C
GC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty