Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Terephthalic acid 99+%

Supplier: Thermo Scientific Chemicals

18072-0250 18072-2500 18072-0010 18072-5000 18072-0051
TS18072-2500EA 29.95 USD InStock
TS18072-2500 TS18072-5000 TS18072-0250 TS18072-0051 TS18072-0010
Terephthalic acid 99+%
Terephthalic acid


Formula: C₈H₆O₄
MW: 166.13 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00002558
CAS Number: 100-21-0
EINECS: 202-830-0
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤0.2 %
Sulfated ash ≤0.001 %
Titration with TBAH ≥99.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty