Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Benzyloxybenzyl alcohol 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (4-(Benzyloxy)phenyl)methanol

18196-0250 18196-0100
TS18196-0250EA 89 USD InStock
TS18196-0250 TS18196-0100
4-Benzyloxybenzyl alcohol 99%
4-Benzyloxybenzyl alcohol


Formula: C₁₄H₁₄O₂
MW: 214.26 g/mol
Melting Pt: 84…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00004654
CAS Number: 836-43-1
EINECS: 212-649-9
Order Now
Specification Test Results
Appearance Off-white Powder
Infrared spectrum Conforms
Melting point 84 to 87 °C
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty