Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(4-Hydroxyphenyl)ethanol 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: p-HPEANSC 59876Tyrosol4-Hydroxyphenethyl alcohol

18250-0050 18250-0250
TS18250-0050EA 76 USD InStock
TS18250-0050 TS18250-0250
2-(4-Hydroxyphenyl)ethanol 98%
2-(4-Hydroxyphenyl)ethanol


Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 89…92 °C
MDL Number: MFCD00002902
CAS Number: 501-94-0
EINECS: 207-930-8
Order Now
Specification Test Results
Appearance Off-white to light beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 89 to 93 °C
GC ≥97.5 %
HPLC ≥90 %
Identification Chlorides: passes test
UV Lambda max 274 nm
UV Lambda max 351 nm
UV Lambda max 525 nm
Absorbance scale 0.75 to 0.83 (A274/A351)
Absorbance scale 0.31 to 0.35 (A525/A351)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty