Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Aminobenzyl alcohol 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-(Hydroxymethyl)aniline(3-Aminophenyl)methanol

18457-0250 18457-0050
TS18457-0050EA 46 USD InStock
TS18457-0050 TS18457-0250
3-Aminobenzyl alcohol 97%
3-Aminobenzyl alcohol


Formula: C₇H₉NO
MW: 123.15 g/mol
Melting Pt: 91…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00007817
CAS Number: 1877-77-6
EINECS: 217-514-8
Order Now
Specification Test Results
Appearance Beige to grey to brown Crystalline powder
Infrared spectrum Conforms
Melting point 91 to 95 °C
GC ≥96.0 %
HPLC ≥98.5 %
Residue after ignition ≤0.1 %
Specific optical rotation +42° to +48° (20 °C, 589 nm) (2 % in 0.1M HCl)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty