Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

9-(Chloromethyl)anthracene 98+%

Supplier: Thermo Scientific Chemicals
Danger

18818-0100 18818-0025
TS18818-0025EA 90 USD InStock
TS18818-0025 TS18818-0100
9-(Chloromethyl)anthracene 98+%
9-(Chloromethyl)anthracene


Formula: C₁₅H₁₁Cl
MW: 226.71 g/mol
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 136…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00001263
CAS Number: 24463-19-2
EINECS: 246-274-7
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance Yellow to golden Powder
Infrared spectrum Conforms
Melting point 136 to 141 °C
HPLC ≥98.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty