Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α,α-Dimethoxytoluene 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Benzaldehyde dimethyl acetal

20979-2500 20979-1000 20979-0025 20979-0010
TS20979-1000EA 48 USD InStock
TS20979-1000 TS20979-0010 TS20979-0025 TS20979-2500
α,α-Dimethoxytoluene 98%
α,α-Dimethoxytoluene


Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 87…89 °C (18 mmHg)
Density: 1.01 g/cm³
Flash Pt: 69 °C
Storage Temperature: Ambient
MDL Number: MFCD00008491
CAS Number: 1125-88-8
EINECS: 214-413-0
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4910 to 1.4940 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty