Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Trifluoromethyl)benzyl alcohol 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-(Hydroxymethyl)benzotrifluoride(4-(Trifluoromethyl)phenyl)methanol

21357-0050
TS21357-0050EA 56 USD InStock
TS21357-0050
4-(Trifluoromethyl)benzyl alcohol 99%
4-(Trifluoromethyl)benzyl alcohol


Formula: C₈H₇F₃O
MW: 176.14 g/mol
Boiling Pt: 78…80 °C (4 mmHg)
Melting Pt: 18…20 °C
Density: 1.28 g/cm³
Flash Pt: 100 °C
MDL Number: MFCD00004661
CAS Number: 349-95-1
EINECS: 206-494-6
Order Now
Specification Test Results
Appearance Clear colorless to light brown Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4590 to 1.4610 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty