Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Aminobenzyl alcohol 98%

Supplier: Thermo Scientific Chemicals
Warning

34824-0500 34824-0050
TS34824-0050EA 115.63 USD InStock
TS34824-0050 TS34824-0500
4-Aminobenzyl alcohol 98%
4-Aminobenzyl alcohol


Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 169…171 °C (11 mmHg)
Melting Pt: 60…63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014782
CAS Number: 623-04-1
EINECS: 210-767-5
Order Now
Specification Test Results
Appearance Off-white to yellow to brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 59 to 65 °C
HPLC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty