Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl-5-hydroxyisophthalate 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Hydroxyisophthalic acid dimethyl esterDimethyl 5-hydroxyisophthalate

35644-0100 35644-0250
TS35644-0100EA 50 USD InStock
TS35644-0100 TS35644-0250
Dimethyl-5-hydroxyisophthalate 99%
Dimethyl 5-hydroxyisophthalate


Formula: C₁₀H₁₀O₅
MW: 210.19 g/mol
Melting Pt: 162…165 °C
MDL Number: MFCD00134367
CAS Number: 13036-02-7
EINECS: 235-899-0
Order Now
Specification Test Results
Appearance White to light beige Powder and/or lumps
Infrared spectrum Conforms
Melting point 162 to 165 °C
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty