Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

2-Hydroxyphenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (2-Hydroxyphenyl)metaboric acid2-Hydroxybenzeneboronic acidDihydroxy(2-hydroxyphenyl)borane

38750-0010 38750-0050
TS38750-0010EA 70 USD InStock
TS38750-0010 TS38750-0050
2-Hydroxyphenylboronic acid 97%
2-Hydroxyphenylboronic acid


Formula: C₆H₇BO₃
MW: 137.93 g/mol
Melting Pt: 173…174 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01074581
CAS Number: 89466-08-0
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty