Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Biphenyl diphenyl ether eutectic

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Dowtherm® ADiphyl

41752-1000 41752-0010
TS41752-1000EA 43 USD InStock
TS41752-1000 TS41752-0010
Biphenyl diphenyl ether eutectic
Biphenyl diphenyl ether eutectic


Formula: C₂₄H₂₀O
MW: 324.423 g/mol
Melting Pt: 13 °C
Density: 1.056 g/cm³
Flash Pt: 123 °C
Storage Temperature: Ambient
MDL Number: MFCD00148859
CAS Number: 8004-13-5
UN: 3082
ADR: 9,III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5885 to 1.5905 (20°C, 589 nm)
GC mix of phenyl ether-biphenyl: approx 70%-30%

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty