Would you like to log in again?
Beakers, New Style, United Scientific Supplies
Supplier: United Scientific Supplies
11115 11116 11110 11117 11112 11114 11111 11113
470339-566EA 25.3 USD InStock
470339-566 470340-016 470339-898 470338-942 470338-920 470339-932 470339-778 470339-042
Beakers, New Style, United Scientific Supplies
Beakers
Polypropylene beakers with excellent clarity and very good chemical resistance.
  • Clear PP
  • Oversized tapered spouts
They are autoclavable and feature raised graduations on the outer surface. New Style beakers feature oversized, tapered spouts for easier pouring of solutions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty