Would you like to log in again?
Centrifuge Tubes, Conical Bottom, PP, HDPE Caps, Sterile, United Scientific Supplies
Supplier: United Scientific Supplies
D1003 D1001 D1003-C D1001-C
470339-992EA 159 USD InStock
470339-992 470340-090 470339-234 470339-406
Centrifuge Tubes, Conical Bottom, PP, HDPE Caps, Sterile, United Scientific Supplies
Tubes Centrifuge Tubes
Pack of 50 racked polypropylene centrifuge tubes with conical bottom. Leak-proof, blue HDPE screw caps are included.
  • Feature silk-screen black graduations (from 2 to 14 ml, at 0.5 ml intervals) and large white marking spot
  • Maximum RCF: 9000
  • Steam sterilized, autoclavable at +121 °C and freezable to –45 °C
Polypropylene tubes include blue HDPE leak-proof screw caps, silk-screen black graduations and a large white marking spot. Available in 15 ml capacity (racked) or 50 ml capacity (bulk in a poly bag).
15 ml tubes are graduated from 2 to 14 ml with graduation intervals of 0.5 ml. 50 ml tubes are graduated from 5 to 47.5 ml with graduation intervals of 2.5 ml.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty