Would you like to log in again?
Self Standing Centrifuge Tubes, PP, HDPE Caps, Sterile, United Scientific Supplies
Supplier: United Scientific Supplies
D1002-C D1002
470339-602EA 168.2 USD InStock
470339-602 470339-388
Self Standing Centrifuge Tubes, PP, HDPE Caps, Sterile, United Scientific Supplies
Tubes Centrifuge Tubes
Sterilized self standing centrifuge tubes.
  • Leak-proof screw
  • Autoclavable at 121 °C and freezable to –45 °C
  • Steam sterilized
  • Maximum RCF 9000
Polypropylene tubes feature a convenient flat bottom so they can stand on their own. Include blue HDPE leak-proof screw caps, silk-screen black graduations and a large white marking spot. Tubes are graduated from 5 to 50 ml with graduation intervals of 2.5 ml.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty