Would you like to log in again?
Centrifuge Tubes, Sterile, United Scientific Supplies
Supplier: United Scientific Supplies
P10406 P10404 P10408
470339-698EA 169.2 USD InStock
470339-698 470340-130 470339-392
Centrifuge Tubes, Sterile, United Scientific Supplies
Tubes Centrifuge Tubes
Leak free centrifuge tubes with conical bottoms.
  • Leak-proof, blue HDPE screw caps included
  • Tubes and caps feature specially-designed flat threads for vacuum-type closure that is leak-free at a pressure of >700 mm Hg
  • Highly smooth hydrophobic internal surfaces minimize wall effect
  • Maximum RCF is 18000 (50 ml)
  • Silk-screen graduations and frosted surface for easy writing
  • Radiation sterilized; DNase-, RNase-, Pyrogen- and latex-free
  • Autoclavable at 121 °C and 15 psi for 30 minutes.
Polypropylene tubes with conical bottoms and blue HDPE leak-proof screw caps included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty