Would you like to log in again?
Reversible Racks for Micro Tubes, PP, United Scientific Supplies
Supplier: United Scientific Supplies
P20206
470339-032EA 84.2 USD InStock
470339-032
Reversible Racks for Micro Tubes, PP, United Scientific Supplies
Racks Tube Racks
Reversible racks with grids for microcentrifuge tubes.
  • Pack of 6 sturdy polypropylene microcentrifuge tube racks
  • Designed with grids on both sides to accommodate tubes 0.5 ml, 1.5 ml and 2.0 ml in capacity
  • Each rack measures 220×150×50 mm and features 96 places in a 12×8 array
  • Positions are coded for easy identification of samples
  • Autoclavable
Pack of six sturdy polypropylene racks for microcentrifuge tubes. Racks contain grids on both sides to accommodate up to 96 microcentrifuge tubes with 0.5, 1.5 or 2.0 ml capacity. Positions are coded for easy identification of samples. Autoclavable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty