Would you like to log in again?
Bottle Top Dispenser, with Bottle, United Scientific Supplies
Supplier: United Scientific Supplies
BTDR-3-SET BTDR-4-SET BTDR-6-SET BTDR-1-SET BTDR-2-SET BTDR-5-SET
470340-238EA 482.15 USD InStock
470340-238 470340-184 470339-690 470339-900 470340-156 470339-390
Bottle Top Dispenser, with Bottle, United Scientific Supplies
Bottle Top Dispensers
Precise dispensing for laboratory solutions, with an included bottle.
  • Includes a bottle
  • Includes adapters to fit a wide range of bottle sizes: 28, 30, 32, 36, 40 and 45 mm
  • All wetted parts made from PTFE and borosilicate glass for maximum chemical resistance
  • Not suitable for use with hydrogen fluoride
  • Individually calibrated and certified in an ISO 17025 accredited laboratory in accordance with ISO 8655 standards
  • Calibration certificate included
Five adapters are included with each unit for use with a wide range of bottle sizes. All wetted parts are made from PTFE and borosilicate glass for maximum chemical resistance. These easy-to-use instruments are suitable for all laboratory applications except HF. Special precautions must be taken for use, cleaning, and storage when working with concentrated fuming acids.
A factory calibration certificate is provided with each bottle top dispenser. Dispensers are easy to disassemble and can be recalibrated in the lab. Fully autoclavable at 121 °C, 15 psi for 10 to 15 minutes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty