Would you like to log in again?
Porcelain Crucible, Wide Form, with Cover, United Scientific Supplies
Supplier: United Scientific Supplies
JCL010
470339-154EA 31.2 USD InStock
470339-154
Porcelain Crucible, Wide Form, with Cover, United Scientific Supplies
Crucibles
Wide form crucibles with a glazed inner and outer surface.
  • Autoclavable
  • Glazed inside and out, except for outside bottom and rim
  • Maximum use temperature of 1150 °C (2102 °F); temperature change rate should not exceed 200 °C (392 °F) per hour
  • Covers included
Excellent resistance to acids and alkalies, not including hydrofluoric acid. Autoclavable and glazed inside and out, except for outside bottom and rim. Designed to withstand temperatures up to 1150 °C; to prevent thermal stress cracks, temperature change rate should no exceed 200 °C per hour.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty