Would you like to log in again?
Storage Vial, PP, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
P60107 P60106
470339-448EA 313 USD InStock
470339-448 470340-192
Storage Vial, PP, United Scientific Supplies
Vials
Polypropylene storage vials for general diagnostics.
  • Polypropylene storage vials
  • Ideal for storage of PCR reagents, enzymes and other diagnostics, biochemical reagents or samples; designed for use with samples from –80 to +121 °C
  • Feature large frosted labeling area, silk-screened graduations and leak-proof HDPE cap
  • DNase, RNase and Pyrogen free
Storage vials are designed for use with samples from –80 to +121 °C. DNase, RNase, and Pyrogen free. Used for storage of PCR reagents, enzymes and other diagnostics, biochemical reagents or samples.
Polypropylene vials feature a large white frosted labeling area, silk screen graduations, and a leak-proof HDPE cap.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty