Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Black Diamond Analog Heating Mantles with Stirrer, United Scientific Supplies

Supplier: United Scientific Supplies
UNAHMTLS-500 UNAHMTLS-1000 UNAHMTLS-250 UNAHMTLS-2000 UNAHMTLS-100 UNAHMTLS-3000 UNAHMTLS-5000 UNAHMTLS-50
470339-500EA 689 USD InStock
470339-500 470339-742 470339-190 470339-298 470339-498 470339-840 470339-724 470339-170
Black Diamond Analog Heating Mantles with Stirrer, United Scientific Supplies
Heaters Heating Mantles
Stir and heat solutions up to 5000 ml.

  • Stainless steel and aluminum construction
  • Glass fiber yarn knitted mantle provides uniform heating to round bottom flasks
  • Separate control knobs for heating and stirring
  • Easy to use and maintain
  • Max. temperature: 400 °C

This series of analog heating mantles includes a built-in stirrer function. Separate knobs control the temperature level up to 400 °C and stirring speed up to 800 rpm. Case is constructed of durable stainless steel and aluminum materials.

Knitted mantle is made from glass fiber yarn and designed to securely hold round bottom flasks with the top half of the flask visible during operation. Available in various sizes to hold flasks from 100 to 5000 ml capacity. Includes a spill collect system to drain solution if a flask breaks during use.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty