Would you like to log in again?
Serological Pipettes, Disposable, Sterile, United Scientific Supplies
Supplier: United Scientific Supplies
UNGD0201-1 UNGD010-1 UNGD0501-1 UNGD1001-1 UNGD5001-1 UNGD2501-1
470338-922EA 44.7 USD InStock
470338-922 470338-930 470339-394 470339-076 470339-330 470339-306
Serological Pipettes, Disposable, Sterile, United Scientific Supplies
Pipettes Serological Pipettes
Disposable Serological pipettes with bi-directional graduations.
  • Individually wrapped sterile pipettes
  • Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink
  • Color coded stripes and packaging enable quick volume identification
  • Gamma sterilized, DNase/RNase/Pyrogen free
Serological pipettes feature black graduations for precise reading and accurate dispensing. Reverse graduations show volume removed or remaining on all pipettes larger than 1 ml. Negative graduations provide additional pipetting volume. Manufactured from transparent USP Class VI polystyrene and sterilized by gamma irradiation. Each lot is DNase, RNase, & Pyrogen free. pipettes are individually wrapped in plastic/paper easy-to-peel packs. Manufactured to ASTM Standard Specification for Serological pipettes E-934.34.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty