Would you like to log in again?
Universal Pipette Tips With Filter, Racked, Sterile, United Scientific Supplies
Supplier: United Scientific Supplies
UNUFT1000UL UNUFT200UL UNUFT10UL UNUFT0300UL UNUFT1250UL UNUFT100UL
470339-710EA 136.6 USD InStock
470339-710 470339-678 470338-898 470340-290 470340-106 470349-754
Universal Pipette Tips With Filter, Racked, Sterile, United Scientific Supplies
Pipette Tips
Racked and sterile universal pipette tips with filter.
  • Manufactured from high grade polypropylene
  • Filter is 100% pure porous polyethylene
  • 99% Bacterial Aerosol Filtration Efficiency (BFE), ASTM F21012 Non-cytotoxic: ISO 10993-5 MRM, Minimum Essential Media Elution, GLP Compliant
  • Non-hemolytic: Modified ASTM F 756-08 Hemolysis Direct Contact GLP with Human Blood
  • DNase, RNase, Pyrogen Free
Protect against cross-contamination using these racked, sterile filter pipette tips. These universal tips are manufactured from polypropylene and the filters are manufactured from polyethylene with a hydrophobic design to prevent liquids and aerosols from entering the body of the micropipet. These tips are non-cytotoxic, non-hemolytic, and have 99.9% Bacterial Aerosol Filtration Efficiency (BFE). DNase, RNase and Pyrogen free. Packaged in racks of 96 with 10 racks in each.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty