Would you like to log in again?
Micropipette Tips, PP, United Scientific Supplies
Supplier: United Scientific Supplies
T1000UL-N T10UL-N T200UL-Y T20UL-N T10ML-N T1000UL-B T5ML-N T200UL-N
470339-456EA 18.29 USD InStock
470339-456 470339-204 470339-618 470340-162 470339-120 470339-814 470340-164 470339-490
Micropipette Tips, PP, United Scientific Supplies
Pipette Tips
Micropipette tips with low liquid retention.
  • Highly smooth, hydrophobic surface provides low liquid retention, increased accuracy and repeatability
  • Inner radial rings enable airtight grip on tip cone
  • Manufactured in a precision molding facility from 100% virgin polypropylene
  • DNase, RNase and Pyrogen free
Manufactured in a precision molding facility from 100% virgin polypropylene, United micropipette tips are DNase, RNase and Pyrogen free. The highly smooth surface is hydrophobic to provide low liquid retention, increased accuracy and repeatability. Inner radial rings enable an airtight grip on the tip cone. Autoclavable at 121 °C and 15 psi for 30 minutes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty