Would you like to log in again?
Universal Low Retention Pipette Tips With Filter, Racked, Sterile, United Scientific Supplies
Supplier: United Scientific Supplies
UNUFT01250UL-LR UNUFT0300UL-LR UNUFT10UL-LR UNUFT100UL-LR UNUFT200UL-LR UNUFT1000UL-LR
470349-802CS 132.2 USD InStock
470349-802 470339-600 470340-014 470340-018 470339-488 470340-200
Universal Low Retention Pipette Tips With Filter, Racked, Sterile, United Scientific Supplies
Pipette Tips
Racked and Sterile low retention universal pipette tips with filter.
  • Manufactured from high grade hydrophobic polypropylene
  • Filter is 100% pure porous polyethylene 99% Bacterial Aerosol Filtration Efficiency (BFE), ASTM F21012
  • Non-cytotoxic: ISO 10993-5 MRM, Minimum Essential Media Elution, GLP Compliant
  • Non-hemolytic: Modified ASTM F 756-08 Hemolysis Direct Contact GLP with Human Blood
  • DNase, RNase, and Pyrogen Free
Protect against cross-contamination using these racked, sterile low retention filter pipette tips. Retention level is 10 times lesser with viscous samples than standard pipette tips. These universal tips are manufactured from polypropylene and the filters are manufactured from polyethylene with a hydrophobic design to prevent liquids and aerosols from entering the body of the micropipet. These tips are non-cytotoxic, non-hemolytic, and have 99.9% Bacterial Aerosol Filtration Efficiency (BFE). DNase, RNase and Pyrogen free. Packaged in racks of 96 with 10 racks in each.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty