Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Aminopyridine 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-PyridylaminePyridin-3-amine

10456-5000 10456-0250 10456-1000
TS10456-5000EA 226 USD InStock
TS10456-5000 TS10456-0250 TS10456-1000
3-Aminopyridine 99%
3-Aminopyridine


Formula: C₅H₆N₂
MW: 94.12 g/mol
Boiling Pt: ∼248 °C (1013 hPa)
Melting Pt: 60…63 °C
Flash Pt: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD00006400
CAS Number: 462-08-8
EINECS: 207-322-2
UN: 2671
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Off-white to brown Crystals or flakes or crystalline mass
Infrared spectrum Conforms
Melting point 60 to 63 °C
GC ≥98.5 %
Moisture ≤3.0 % (as packaged)
Viscosity 0.2 to 0.6 s
Free acid (CH3COOH) ≤0.06 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty